Bismuth(III) nitrate pentahydrate ≥99.999% (Bi basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Bismuth trinitrate pentahydrate

93-8303-5G 93-8303-25G 93-8303-100G
100204-452EA 52.37 USD
100204-452 100209-232 100209-230
Bismuth(III) nitrate pentahydrate ≥99.999% (Bi basis), PURATREM
Bismuth(III) nitrate pentahydrate
Formula: Bi(NO₃)₃·5H₂O
MW: 485.07 g/mol
Boiling Pt: 75…80 °C (1013 hPa)
Melting Pt: 30 °C
Density: 2.736 g/cm³ (20 °C)
MDL Number: MFCD00149157
CAS Number: 10035-06-0
EINECS: 233-791-8
UN: 1477
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR