Lead tetraacetate ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

93-8270-50G 93-8270-250G
100196-838EA 101.91 USD
100196-838 100202-730
Lead tetraacetate ≥95%
Lead tetraacetate
Formula: (H₃CCOO)₄Pb
Melting Pt: 175 °C
Density: 2.28
Storage Temperature: Ambient
MDL Number: MFCD00008693
CAS Number: 546-67-8
EINECS: 208-908-0
UN: 1616
ADR: 6.1,III
Merck Index: 13,05442

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR