Lead selenide ≥99.99% (Pb basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Lead(II) selenide

93-8257-25G 93-8257-5G
100197-970EA 489.72 USD
100197-970 100197-972
Lead selenide ≥99.99% (Pb basis), PURATREM
Lead selenide
Formula: PbSe
MW: 286.16
Melting Pt: 1065 °C
Density: 8.10
MDL Number: MFCD00016273
CAS Number: 12069-00-0
EINECS: 235-109-4
UN: 3283
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR