Lead(II) acetate trihydrate ≥99% ACS

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate

93-8250-1000G 93-8250-250G
100208-604EA 226.46 USD
100208-604 100203-602
Lead(II) acetate trihydrate ≥99% ACS
Lead(II) acetate trihydrate
Formula: (H₃CCOO)₂Pb·3H₂O
MW: 379.34 g/mol
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 75 °C (dec.)
Density: 2.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150023
CAS Number: 6080-56-4
EINECS: 206-104-4
UN: 1616
ADR: 6.1,III
Merck Index: 13,05415
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=351511a44fbb598a2d7fa1fa4ab395427b6168f1&vid=67e858e5f0dc4d04dd796541b3c0ad5aa73f8dc8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR