Lead(II) sulphide ≥99.999% (Pb basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Lead sulphide
93-8233-25G
93-8233-5G
100209-462EA
723.26
USD
100209-462
100204-446
Lead(II) sulphide ≥99.999% (Pb basis), PURATREM
Lead(II) sulphide
Formula:
̍PbS Boiling Pt: 1344 °C (1013 hPa) Melting Pt: 1114 °C Density: 7.5 g/cm³ (20 °C) |
MDL Number:
MFCD00016280 CAS Number: 1314-87-0 EINECS: 215-246-6 UN: 3077 ADR: 9,III Merck Index: 12,05445 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...