Lead(II) sulfate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-8232-1000G 93-8232-250G
100203-600EA 496.8 USD
100203-600 100204-444
Lead(II) sulfate ≥99%
Lead(II) sulphate
Formula: O₄PbS
Melting Pt: 1170 °C
Density: 6.0631 g/cm³ (18 °C)
MDL Number: MFCD00011166
CAS Number: 7446-14-2
EINECS: 231-198-9
UN: 2291
ADR: 6.1,III
Merck Index: 12,05444

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR