Lead(IV) oxide ≥97% ACS

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Lead dioxide

93-8210-100G 93-8210-500G
100202-316EA 100.49 USD
100202-316 100205-906
Lead(IV) oxide ≥97% ACS
Lead(IV) oxide
Formula: O₂Pb
MW: 239.2 g/mol
Melting Pt: 290 °C (dec.)
Density: 9.4 g/cm³ (20 °C)
MDL Number: MFCD00011165
CAS Number: 1309-60-0
EINECS: 215-174-5
UN: 1872
ADR: 5.1,III
Merck Index: 12,05424

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR