Dithallium trioxide ≥99.5% (Tl basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-8117-25G 93-8117-100G
100211-304EA 210.89 USD
100211-304 100207-318
Dithallium trioxide ≥99.5% (Tl basis)
Dithallium trioxide
Formula: O₃Tl₂
Melting Pt: ∼717 °C
Density: 5.56 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011276
CAS Number: 1314-32-5
EINECS: 215-229-3
UN: 1707
ADR: 6.1,II
Merck Index: 12,09404

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR