Thallium(I) nitrate ≥99.5% (Tl basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Thallium nitrate
93-8109-100G
93-8109-25G
100208-236EA
754.4
USD
100208-236
100208-238
Thallium(I) nitrate ≥99.5% (Tl basis)
Thallium(I) nitrate
Formula:
NO₃Tl MW: 266.39 g/mol Boiling Pt: 430 °C Melting Pt: 206 °C Density: 1.273 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011280 CAS Number: 10102-45-1 EINECS: 233-273-1 UN: 2727 ADR: 6.1,II Merck Index: 12,09400 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...