Thallium(I) ethanolate ≥95%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Thallium (I) ethoxide
93-8105-5G
93-8105-25G
100210-572EA
165.6
USD
100210-572
100209-222
Thallium(I) ethanolate ≥95%
Thallium(I) ethanolate
Alkoxide, Metal Alkoxides, Volatile Precursors for CVD
Formula:
C₂H₅OTl MW: 249.44 g/mol Boiling Pt: 130 °C (dec.) Melting Pt: –3 °C Density: 3.52 g/cm³ (20 °C) Flash Pt: 60 °C |
MDL Number:
MFCD00009072 CAS Number: 20398-06-5 EINECS: 243-786-2 UN: 1707 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...