Mercury ≥99.999% ACS

Supplier: STREM CHEMICALS INC MS
Danger

93-8046-2250G 93-8046-450G
100210-768EA 2341.05 USD
100210-768 100198-058
Mercury ≥99.999% ACS
Mercury
Formula: Hg
MW: 200.59 g/mol
Boiling Pt: 356.5 °C (1013 hPa)
Melting Pt: -39 °C
Density: 13.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011035
CAS Number: 7439-97-6
EINECS: 231-106-7
UN: 2809
ADR: 8,III
Merck Index: 13,05925

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR