Mercury(II) cyanide ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Mercury dicyanide, Mercuric cyanide

93-8041-50G 93-8041-250G
100200-212EA 409.05 USD
100200-212 100200-210
Mercury(II) cyanide ≥99%
Mercury(II) cyanide
Formula: C₂HgN₂
MW: 252.63
Melting Pt: 3.77 Dec.
Density: 3.77
Storage Temperature: Ambient
MDL Number: MFCD00011037
CAS Number: 592-04-1
EINECS: 209-741-6
UN: 1636
ADR: 6.1 II
Merck Index: 12,05928

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR