Mercury(II) sulfate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Mercury sulphate, Mercuric sulphate

93-8036-250G 93-8036-50G
100208-234EA 380.74 USD
100208-234 100202-308
Mercury(II) sulfate ≥98%
Mercury(II) sulphate
Formula: HgSO₄
MW: 296.65 g/mol
Melting Pt: 850 °C
Density: 6.47 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011047
CAS Number: 7783-35-9
EINECS: 231-992-5
UN: 1645
ADR: 6.1,II
Merck Index: 13,05914

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR