Mercury(I) nitrate monohydrate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Dimercury dinitrate monohydrate, Mercurous nitrate monohydrate, Mercury(I) nitrate hydrate

93-8020-250G 93-8020-50G
100210-566EA 448.68 USD
100210-566 100210-568
Mercury(I) nitrate monohydrate ≥98%
Mercury(I) nitrate monohydrate
Formula: Hg₂N₂O₆·1H₂O
MW: 543.21 g/mol
Melting Pt: 70 °C
Density: 4.79 g/cm³ (20 °C)
MDL Number: MFCD00150813
CAS Number: 7782-86-7
EINECS: 233-886-4
UN: 1627
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR