Mercury selenide ≥99.99% (Hg basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

93-8013-1G 93-8013-25G 93-8013-5G
100208-602EA 62.28 USD
100208-602 100206-168 100198-462
Mercury selenide ≥99.99% (Hg basis), PURATREM
Mercury selenide
Formula: HgSe
MW: 279.55 g/mol
Melting Pt: 998 °C
Density: 7.87 g/cm³ (25 °C)
MDL Number: MFCD00049743
CAS Number: 20601-83-6
EINECS: 243-910-5
UN: 2025
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR