Mercury(II) iodide ≥99% ACS
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Mercury diiodide, Mercuric iodide
93-8008-250G
93-8008-50G
100202-300EA
389.23
USD
100202-300
100204-430
Mercury(II) iodide ≥99% ACS
Mercury(II) iodide
Formula:
HgI₂ MW: 454.4 g/mol Boiling Pt: 354 Melting Pt: 259 Density: 6.271 Storage Temperature: Ambient |
MDL Number:
MFCD00011044 CAS Number: 7774-29-0 EINECS: 231-873-8 UN: 1638 ADR: 6.1 II Merck Index: 13,05905 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...