Mercury(II) chloride ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-8004-50G 93-8004-250G
100204-428EA 110.4 USD
100204-428 100210-766
Mercury(II) chloride ≥99%
Mercury(II) chloride
Formula: HgCl₂
Boiling Pt: 302 °C
Melting Pt: 276 °C
Density: 5.44
Storage Temperature: Ambient
MDL Number: MFCD00011041
CAS Number: 7487-94-7
EINECS: 231-299-8
UN: 1624
ADR: 6.1,II
Merck Index: 13,05901

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR