Mercury(II) bromide ACS
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Mercuric bromide, Mercury dibromide
93-8003-250G
93-8003-50G
100208-600EA
745.91
USD
100208-600
100200-608
Mercury(II) bromide ACS
Mercury(II) bromide
Formula:
Br₂Hg MW: 360.4 g/mol Boiling Pt: 320 Melting Pt: 236 Density: 6.109 Storage Temperature: Ambient |
MDL Number:
MFCD00011040 CAS Number: 7789-47-1 EINECS: 232-169-3 UN: 1634 ADR: 6.1 II Merck Index: 13,05900 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...