Gold trichloride ≥99% (99.9%-Au)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Gold(III) chloride, Gold (III) chloride
93-7907-5G
93-7907-1G
100207-174EA
769.97
USD
100207-174
100207-172
Gold trichloride ≥99% (99.9%-Au)
Gold trichloride
Formula:
AuCl₃ MW: 303.32 Melting Pt: 254 °C (dec.) Density: 4.3 g/cm³ (20 °C) |
MDL Number:
MFCD00014172 CAS Number: 13453-07-1 EINECS: 236-623-1 UN: 3260 ADR: 8 III Merck Index: 13,04536 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...