Gold(III) oxide ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-7903-1G 93-7903-5G
100198-054EA 318.46 USD
100198-054 100202-728
Gold(III) oxide ≥99%
Gold(III) oxide
Formula: Au₂O₃
MW: 441.93
Melting Pt: 150 (dec.)
Density: 3.6 g/cm³ (20 °C)
MDL Number: MFCD00014173
CAS Number: 1303-58-8
EINECS: 215-122-1
Merck Index: 12,04545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR