Platinum ≥99.9%, powder

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Platinium

93-7833-1G 93-7833-5G
100197-962EA 358.09 USD
100197-962 100200-606
Platinum ≥99.9%, powder
Platinum
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.45
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 3089
ADR: 4.1 III
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR