Sodium tetrachloroplatinate(II) hydrate
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Disodium tetrachloroplatinate hydrate, Sodium platinum chloride hydrate, Sodium chloroplatinate(II) hydrate
93-7810-5G
93-7810-1G
100207-316EA
632.68
USD
100207-316
100199-190
Sodium tetrachloroplatinate(II) hydrate
Sodium tetrachloroplatinate(II) hydrate
Formula:
Na₂PtCl₄·xH₂O MW: 382.88 g/mol (anhy) Melting Pt: 100 °C |
MDL Number:
MFCD00150188 CAS Number: 207683-21-4 EINECS: 233-051-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...