Platinum(II) iodide 98
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Platinum diiodide, Platinum(II) iodide
93-7808-5G
93-7808-1G
100202-472EA
1197.42
USD
100202-472
100205-770
Platinum(II) iodide 98
Platinum(II) iodide
Formula:
I₂Pt MW: 448.89 Melting Pt: 360 (dec.) Density: 6.403 g/cm³ (25 °C) |
MDL Number:
MFCD00011183 CAS Number: 7790-39-8 EINECS: 232-204-2 Merck Index: 12,07688 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...