Diammonium tetrachloroplatinate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Ammonium tetrachloroplatinate(II), Ammonium tetrachloroplatinate
93-7802-5G
93-7802-1G
100207-498EA
772.8
USD
100207-498
100199-664
Diammonium tetrachloroplatinate ≥99%
Diammonium tetrachloroplatinate
Formula:
H₈Cl₄N₂Pt MW: 372.97 g/mol Melting Pt: 140 °C Density: 2.93 g/cm³ (20 °C) |
MDL Number:
MFCD00010885 CAS Number: 13820-41-2 EINECS: 237-499-1 UN: 3288 ADR: 6.1,III Merck Index: 12,00583 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...