Ammonium hexachloroplatinate(IV) ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-7801-1G
93-7801-5G
100202-580EA
417.54
USD
100202-580
100208-734
Ammonium hexachloroplatinate(IV) ≥99%
Ammonium hexachloroplatinate(IV)
Formula:
H₈Cl₆N₂Pt Melting Pt: Dec. Density: 3.06 g/cm³ (20 °C) |
MDL Number:
MFCD00010886 CAS Number: 16919-58-7 EINECS: 240-973-0 UN: 3288 ADR: 6.1,III Merck Index: 12,00582 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...