Iridium(IV) oxide ≥99% (99.9+%-Ir)

Supplier: STREM CHEMICALS INC MS
Warning

93-7702-1G 93-7702-5G
100207-168EA 711.94 USD
100207-168 100207-170
Iridium(IV) oxide ≥99% (99.9+%-Ir)
Iridium(IV) oxide
Formula: IrO₂
Boiling Pt: –27 °C (1013 hPa)
Melting Pt: 1100 °C (dec.)
Density: 3.09 g/cm³ (18 °C)
MDL Number: MFCD00011065
CAS Number: 12030-49-8
EINECS: 234-743-9
UN: 1479
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR