Disodium hexachloroosmate hydrate

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Sodium hexachloroosmate(IV) hydrate

93-7612-2G 93-7612-500MG
100202-298EA 576.06 USD
100202-298 100202-296
Disodium hexachloroosmate hydrate
Disodium hexachloroosmate hydrate
Formula: Na₂OsCl₆·xH₂O
MW: 448.93 g/mol (anhy)
MDL Number: MFCD00150185
CAS Number: 207683-17-8
EINECS: 215-152-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR