Rhenium pentachloride ≥99.9% (Re basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-7506-1G 93-7506-5G
100200-798EA 233.54 USD
100200-798 100195-928
Rhenium pentachloride ≥99.9% (Re basis)
Rhenium pentachloride
Formula: Cl₅Re
MW: 363.47 g/mol
Melting Pt: Dec.
Density: 4,9
MDL Number: MFCD00011197
CAS Number: 13596-35-5
EINECS: 237-042-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR