Dirhenium heptaoxide ≥99.99% (Re basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Rhenium (VII) oxide

93-7505-5G 93-7505-1G
100203-584EA 476.98 USD
100203-584 100202-294
Dirhenium heptaoxide ≥99.99% (Re basis), PURATREM
Dirhenium heptaoxide
Formula: O₇Re₂
MW: 484.41 g/mol
Boiling Pt: 363 °C (1013 hPa)
Melting Pt: 220 °C
Density: 6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011199
CAS Number: 1314-68-7
EINECS: 215-241-9
UN: 1479
ADR: 5.1,II
Merck Index: 12,08344

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR