Dipotassium hexachlororhenate ≥99.9% (Re basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Potassium hexachlororhenate(IV)
93-7502-1G
93-7502-5G
100202-290EA
206.65
USD
100202-290
100202-292
Dipotassium hexachlororhenate ≥99.9% (Re basis)
Dipotassium hexachlororhenate
Formula:
Cl₆K₂Re MW: 477.12 g/mol Density: 3.34 |
MDL Number:
MFCD00058746 CAS Number: 16940-97-9 EINECS: 241-008-6 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...