Tungsten ≥99.95%, wire

Supplier: STREM CHEMICALS INC MS
Warning

93-7438-50MT 93-7438-250MT
100202-724EA 150.03 USD
100202-724 100208-732
Tungsten ≥99.95%, wire
Formula: W
Boiling Pt: 5660 °C
Melting Pt: 3410 °C
Density: 19.3
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR