Tungsten disulphide ≥99.5% (W basis)

Supplier: STREM CHEMICALS INC MS

Synonyms: Tungsten sulphide, Tungsten (IV) sulphide

93-7424-50G 93-7424-250G
100199-662EA 114.65 USD
100199-662 100207-158
Tungsten disulphide ≥99.5% (W basis)
Tungsten disulphide
Formula: S₂W
MW: 247.97
Melting Pt: 1250 °C (dec.)
Density: 7.5
MDL Number: MFCD00011467
CAS Number: 12138-09-9
EINECS: 235-243-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR