Tungsten trioxide ≥99.99% (W basis), sintered lumps, PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Tungsten (VI) oxide
93-7411-10G
93-7411-50G
100197-956EA
69.35
USD
100197-956
100197-958
Tungsten trioxide ≥99.99% (W basis), sintered lumps, PURATREM
Tungsten trioxide
Formula:
O₃W MW: 231.84 Melting Pt: 1473 Density: 7.16 |
MDL Number:
MFCD00011466 CAS Number: 1314-35-8 EINECS: 215-231-4 Merck Index: 12,09947 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...