Phosphotungstic acid hydrate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Phosphotungstic acid hydrate, Wolframophosphoric acid hydrate, Dodecawolframophosphoric acid hydrate, 12-Tungstophosphate hydrate, Tungstophosphoric Acid Hydrate
93-7402-25G
93-7402-500G
93-7402-100G
100210-552EA
65.11
USD
100210-552
100200-198
100202-576
Phosphotungstic acid hydrate ≥99%
Phosphotungstic acid hydrate
Formula:
H₃[P(W₃O₁₀)₄]·nH₂O MW: 2898.07 g/mol Melting Pt: 107 °C |
MDL Number:
MFCD00011342 CAS Number: 12501-23-4 EINECS: 215-682-7 UN: 3260 ADR: 8,III Merck Index: 13,07450 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...