Tantalum ≥99.98% (Nb-25ppm), Powder
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-7332-10G
93-7332-50G
100199-182EA
92
USD
100199-182
100202-470
Tantalum ≥99.98% (Nb-25ppm), Powder
Tantalum
Formula:
Ta Boiling Pt: 5425 °C Melting Pt: 2996 °C Density: 16.6 g/cm³ (20 °C) |
MDL Number:
MFCD00011252 CAS Number: 7440-25-7 EINECS: 231-135-5 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...