Tantalum pentafluoride 99,5

Supplier: STREM CHEMICALS INC MS
Danger

93-7325-5G 93-7325-25G
100203-582EA 157.11 USD
100203-582 100200-194
Tantalum pentafluoride 99,5
Tantalum pentafluoride

Halides for CVD & ALD

Formula: F₅Ta
MW: 275.94
Boiling Pt: 229
Melting Pt: 97
Density: 4.74
MDL Number: MFCD00042542
CAS Number: 7783-71-3
EINECS: 232-022-3
UN: 1759
ADR: 8 II
Merck Index: 12,09225

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR