Tantalum pentachloride ≥99.99% (Ta basis), PURATREM, resublimed

Supplier: STREM CHEMICALS INC MS
Danger

93-7324-10G 93-7324-50G
100208-594EA 100.49 USD
100208-594 100201-144
Tantalum pentachloride ≥99.99% (Ta basis), PURATREM, resublimed
Tantalum pentachloride
Formula: Cl₅Ta
Boiling Pt: 242 °C
Melting Pt: 144 °C (subl.)
Density: 3.68 g/cm³ (20 °C)
MDL Number: MFCD00011253
CAS Number: 7721-01-9
EINECS: 231-755-6
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR