Lutetium trinitrate hydrate ≥99.9% (Lu basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Lutetium (III) nitrate hydrate
93-7113-5G
93-7113-1G
100202-280EA
246.28
USD
100202-280
100210-548
Lutetium trinitrate hydrate ≥99.9% (Lu basis)
Lutetium trinitrate hydrate
Formula:
LuN₃O₉·H₂O MW: 379 |
MDL Number:
MFCD00150421 CAS Number: 100641-16-5 EINECS: 233-241-7 UN: 1477 ADR: 5.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...