Lutetium trichloride, anhydrous ≥99.9% (Lu basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-7101-5G
93-7101-1G
100197-952EA
495.38
USD
100197-952
100197-950
Lutetium trichloride, anhydrous ≥99.9% (Lu basis)
Lutetium trichloride
Formula:
Cl₃Lu MW: 281.33 g/mol Melting Pt: 905 °C Density: 3.98 g/cm³ (25 °C) |
MDL Number:
MFCD00011099 CAS Number: 10099-66-8 EINECS: 233-240-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...