Ytterbium ≥99.9%, ingot
Supplier: STREM CHEMICALS INC MS
93-7032-25G
93-7032-5G
100196-998EA
823.75
USD
100196-998
100202-276
Ytterbium ≥99.9%, ingot
Ytterbium
Formula:
Yb Boiling Pt: 1427 °C Melting Pt: 824 °C Density: 6.977 g/cm³ (20 °C) |
MDL Number:
MFCD00011286 CAS Number: 7440-64-4 EINECS: 231-173-2 Merck Index: 12,10239 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...