Ytterbium ≥99.9%, chips

Supplier: STREM CHEMICALS INC MS

93-7031-5G 93-7031-25G
100201-142EA 127.38 USD
100201-142 100203-576
Ytterbium ≥99.9%, chips
Ytterbium
Formula: Yb
Boiling Pt: 1427 °C
Melting Pt: 824 °C
Density: 6.977 g/cm³ (20 °C)
MDL Number: MFCD00011286
CAS Number: 7440-64-4
EINECS: 231-173-2
Merck Index: 12,10239

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR