Ytterbium(III) chloride, anhydrous ≥99.9% (Yb basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-7025-5G 93-7025-25G 93-7025-1G
100199-660EA 157.11 USD
100199-660 100199-658 100207-146
Ytterbium(III) chloride, anhydrous ≥99.9% (Yb basis)
Ytterbium(III) chloride
Formula: Cl₃Yb
MW: 279.4
Melting Pt: 860
MDL Number: MFCD00011288
CAS Number: 10361-91-8
EINECS: 233-800-5
Merck Index: 13,10014

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR