Ytterbium(III) oxide ≥99.9% (Yb basis)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Diytterbium trioxide, Ytterbium oxide
93-7008-100G
93-7008-25G
100197-948EA
261.85
USD
100197-948
100209-208
Ytterbium(III) oxide ≥99.9% (Yb basis)
Ytterbium(III) oxide
Formula:
O₃Yb₂ MW: 394.08 g/mol Melting Pt: 2227 °C Density: 9.2 g/cm³ (20 °C) |
MDL Number:
MFCD00011290 CAS Number: 1314-37-0 EINECS: 215-234-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...