Ytterbium trinitrate pentahydrate ≥99.9% (Yb basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Ytterbium (III) nitrate pentahydrate
93-7006-5G
93-7006-25G
100210-542EA
35.38
USD
100210-542
100200-794
Ytterbium trinitrate pentahydrate ≥99.9% (Yb basis)
Ytterbium trinitrate pentahydrate
Formula:
N₃O₉Yb·5H₂O MW: 449.13 g/mol |
MDL Number:
MFCD00149883 CAS Number: 35725-34-9 EINECS: 237-384-6 UN: 1477 ADR: 5.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...