Ytterbium(III) fluoride ≥99.9% (Yb basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-7004-5G 93-7004-25G
100204-418EA 106.15 USD
100204-418 100202-272
Ytterbium(III) fluoride ≥99.9% (Yb basis)
Ytterbium(III) fluoride
Formula: F₃Yb
Boiling Pt: 2200 °C
Melting Pt: 1157 °C
Density: 8.168
MDL Number: MFCD00049615
CAS Number: 13760-80-0
EINECS: 237-354-2
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR