Ytterbium(III) chloride hexahydrate ≥99.9% (Yb basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-7003-10G
93-7003-50G
100196-818EA
63.69
USD
100196-818
100211-288
Ytterbium(III) chloride hexahydrate ≥99.9% (Yb basis)
Ytterbium(III) chloride hexahydrate
Formula:
Cl₃Yb·6H₂O MW: 387.49 g/mol Melting Pt: 154 °C Density: 2.57 g/cm³ (20 °C) |
MDL Number:
MFCD00149885 CAS Number: 10035-01-5 EINECS: 233-800-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...