Thulium ≥99.9%, foil
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-6918-50X50MM
93-6918-25X25MM
100207-312EA
4169.72
USD
100207-312
100196-996
Thulium ≥99.9%, foil
Thulium
Formula:
Tm MW: 168.93 g/mol Boiling Pt: 1727 °C Melting Pt: 1545 °C Density: 9.332 |
MDL Number:
MFCD00011281 CAS Number: 7440-30-4 EINECS: 231-140-2 UN: 3178 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...