Thulium ≥99.9%, powder
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-6915-5G
93-6915-1G
100198-446EA
884.62
USD
100198-446
100198-444
Thulium ≥99.9%, powder
Thulium
Formula:
Tm Boiling Pt: 1727 °C (1013 hPa) Melting Pt: 1544.85 °C Density: 9.318 g/cm³ (25 °C) |
MDL Number:
MFCD00011281 CAS Number: 7440-30-4 EINECS: 231-140-2 UN: 3089 ADR: 4,III Merck Index: 12,09535 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...