Erbium ≥99.9%, foil
Supplier: STREM CHEMICALS INC MS
93-6834-50X50MM
93-6834-25X25MM
100199-166EA
1538.52
USD
100199-166
100199-168
Erbium ≥99.9%, foil
Erbium
Formula:
Er MW: 167.26 g/mol Boiling Pt: 2863 °C Melting Pt: 1529 °C Density: 9,006 |
MDL Number:
MFCD00010987 CAS Number: 7440-52-0 EINECS: 231-160-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...