Erbium ≥99.9%, ingot

Supplier: STREM CHEMICALS INC MS

93-6832-10G 93-6832-50G
100205-740EA 222.22 USD
100205-740 100207-494
Erbium ≥99.9%, ingot
Erbium
Formula: Er
MW: 167.26 g/mol
Boiling Pt: 2863 °C
Melting Pt: 1529 °C
Density: 9,006
MDL Number: MFCD00010987
CAS Number: 7440-52-0
EINECS: 231-160-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR